Human Recombinant Ube2I
Supplier: Bon Opus Biosciences
CF30-10UG CF30-1MG CF30-500UG CF30-50UG
102864-686EA 50.62 USD
102864-686 102864-688 102864-690 102864-692
Human Recombinant Ube2I
Enzymes
It offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products.
  • Consistent sizing options
  • Datasheet included
  • High purity, low endotoxin levels
Recombinant human ubiquitin-conjugating enzyme E2 I is produced by our E.coli expression system and the target gene encoding Met1-Ser158 is expressed with a GST tag at the N-terminus.
Información accesorios: Accessories not included.
Información de suministro: Shipped via FedEx.
Envase: Shipped domestic on polar packs in a 9×6×7" box, or shipped internationally on dry ice in a 14" cube box.
Precaución: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only. The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.