Recombinant Human Tpm3, Bon Opus Biosciences
Supplier: Bon Opus Biosciences
CF21-10UG CF21-1MG CF21-500UG CF21-50UG
102864-630EA 118.46 USD
102864-630 102864-632 102864-634 102864-636
Recombinant Human Tpm3, Bon Opus Biosciences
Proteins and Peptides
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels
Recombinant Human Tropomyosin alpha-3 is produced by our E.coli expression system and the target gene encoding Met1-Met248 is expressed.
Información accesorios: Accessories not included.
Información de suministro: Shipped Via FedEx
Envase: Shipped at ambient temperature via FedEx envelope.
Precaución: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.