Human Recombinant Dck
Supplier: Bon Opus Biosciences
CF06-10UG CF06-1MG CF06-500UG CF06-50UG
102864-518EA 107.69 USD
102864-518 102864-520 102864-522 102864-524
Human Recombinant Dck
Enzymes
It offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products.
  • Consistent sizing options
  • Datasheet included
  • High purity, low endotoxin levels
Recombinant human deoxycytidine kinase is produced by our E.coli expression system and the target gene encoding Met1-Leu260 is expressed with a 6His, T7 tag at the N-terminus.
Información accesorios: Accessories not included.
Información de suministro: Shipped via FedEx.
Envase: Shipped domestic on polar packs in a 9×6×7" box, or shipped internationally on dry ice in a 14" cube box.
Precaución: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only. The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.