Recombinant Human Bcas2, Bon Opus Biosciences
Supplier: Bon Opus Biosciences
CE68-10UG CE68-1MG CE68-500UG CE68-50UG
102864-262EA 170.15 USD
102864-262 102864-264 102864-266 102864-268
Recombinant Human Bcas2, Bon Opus Biosciences
Proteins and Peptides
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels
Recombinant Human BCAS2 is produced by our E.coli expression system and the target gene encoding Ala2-Phe225 is expressed with a T7 tag at the N-terminus, 6His tag at the C-terminus.
Información accesorios: Accessories not included.
Información de suministro: Shipped Via FedEx
Envase: Shipped at ambient temperature via FedEx envelope.
Precaución: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.