Recombinant Human Znf75A, Bon Opus Biosciences
Supplier: Bon Opus Biosciences
CE51-10UG CE51-1MG CE51-500UG CE51-50UG
102864-158EA 170.15 USD
102864-158 102864-160 102864-162 102864-164
Recombinant Human Znf75A, Bon Opus Biosciences
Proteins and Peptides
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels
Recombinant Human Zinc Finger Protein 75A is produced by our E.coli expression system and the target gene encoding Ser58-Lys162 is expressed with a 6His tag at the N-terminus.
Información accesorios: Accessories not included.
Información de suministro: Shipped Via FedEx
Envase: Shipped at ambient temperature via FedEx envelope.
Precaución: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.