Human Recombinant Wbp2
Supplier: Bon Opus Biosciences
CE22-10UG CE22-1MG CE22-500UG CE22-50UG
102863-934EA 170.15 USD
102863-934 102863-936 102863-938 102863-940
Human Recombinant Wbp2
Enzymes
It offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent sizing options
  • Datasheet included
  • High purity, low endotoxin levels
Recombinant human WW domain-binding protein 2 is produced by our E.coli expression system and the target gene encoding Met1-Ala100 is expressed with a 6His tag at the N-terminus.
Información accesorios: Accessories not included.
Información de suministro: Shipped via FedEx.
Envase: Shipped at ambient temperature via FedEx envelope.
Precaución: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only. The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.