Human Recombinant TIM-3
Supplier: Bon Opus Biosciences
CX71-10UG CX71-1MG CX71-500UG CX71-50UG
102509-372EA 79.69 USD
102509-372 102509-374 102509-376 102509-378
Human Recombinant TIM-3
Proteins and Peptides
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels
Recombinant Human TIM-3 is produced by our Mammalian expression system and the target gene encoding Ser22-Arg200 is expressed with a Fc, 6His tag at the C-terminus.
Información accesorios: Accessories not included.
Información de suministro: Shipped Via FedEx
Envase: Shipped at ambient temperature via FedEx envelope.
Precaución: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.