Human Recombinant V-set and immunoglobulin domain-containing protein 4
Supplier: Bon Opus Biosciences
CX69-10UG CX69-1MG CX69-500UG CX69-50UG
102498-164EA 79.69 USD
102498-164 102498-166 102498-168 102509-362
Human Recombinant V-set and immunoglobulin domain-containing protein 4
Proteins and Peptides
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels
Recombinant Human VSIG4 is produced by our Mammalian expression system and the target gene encoding Arg20-Val284 is expressed with a Fc tag at the C-terminus.
Información accesorios: Accessories not included.
Información de suministro: Shipped Via FedEx
Envase: Shipped at ambient temperature via FedEx envelope.
Precaución: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.