Mouse Recombinant Ephrin-A5
Supplier: Bon Opus Biosciences
CX23-10UG CX23-1MG CX23-500UG CX23-50UG
102497-900EA 107.69 USD
102497-900 102497-902 102497-904 102497-906
Mouse Recombinant Ephrin-A5
Proteins and Peptides
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels
Recombinant Mouse Ephrin-A5 is produced by our Mammalian expression system and the target gene encoding Gln21-Gln206 is expressed with a 6His tag at the C-terminus.
Información accesorios: Accessories not included.
Información de suministro: Shipped Via FedEx
Envase: Shipped at ambient temperature via FedEx envelope.
Precaución: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.