Human Recombinant Interferon Alpha/Beta Receptor 2
Supplier: Bon Opus Biosciences
CX10-10UG CX10-1MG CX10-500UG CX10-50UG
102497-820EA 107.69 USD
102497-820 102497-822 102497-824 102497-826
Human Recombinant Interferon Alpha/Beta Receptor 2
Proteins and Peptides
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels
Recombinant Human Interferon alpha/beta Receptor 2 is produced by our Mammalian expression system and the target gene encoding Ile27-Lys243 is expressed with a Fc tag at the C-terminus.
Información accesorios: Accessories not included.
Información de suministro: Shipped Via FedEx
Envase: Shipped at ambient temperature via FedEx envelope.
Precaución: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.