Human Recombinant Psg1 Cc66
Supplier: Bon Opus Biosciences
CC66-10UG CC66-1MG CC66-500UG CC66-50UG
102497-580EA 170.15 USD
102497-580 102497-582 102497-584 102497-586
Human Recombinant Psg1 Cc66
Enzymes
It offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products.
  • Consistent sizing options
  • Datasheet included
  • High purity, low endotoxin levels
Recombinant human pregnancy-specific beta-1-glycoprotein 1 is produced by our mammalian expression system and the target gene encoding Gln35-Pro419 is expressed with a 6His tag at the C-terminus.
Información accesorios: Accessories not included.
Información de suministro: Shipped via FedEx.
Envase: Shipped at ambient temperature via FedEx envelope.
Precaución: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only. The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.