Human Recombinant Semaphorin-4B
Supplier: Bon Opus Biosciences
CC34-10UG CC34-1MG CC34-500UG CC34-50UG
102509-274EA 170.15 USD
102509-274 102509-276 102509-278 102509-280
Human Recombinant Semaphorin-4B
Proteins and Peptides
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels
Recombinant Human Semaphorin 4B is produced by our Mammalian expression system and the target gene encoding Leu39-Glu712 is expressed with a Fc, 6His tag at the C-terminus.
Información accesorios: Accessories not included.
Información de suministro: Shipped Via FedEx
Envase: Shipped at ambient temperature via FedEx envelope.
Precaución: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.