Human Recombinant Protein CYR61
Supplier: Bon Opus Biosciences
CB98-10UG CB98-1MG CB98-500UG CB98-50UG
102497-344EA 107.69 USD
102497-344 102497-346 102497-348 102497-350
Human Recombinant Protein CYR61
Proteins and Peptides
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels
Recombinant Human CCN1 is produced by our Mammalian expression system and the target gene encoding Thr25-Asp381 is expressed with a Fc tag at the C-terminus.
Información accesorios: Accessories not included.
Información de suministro: Shipped Via FedEx
Envase: Shipped at ambient temperature via FedEx envelope.
Precaución: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.