Human Recombinant Activin Receptor Type-1
Supplier: Bon Opus Biosciences
CA76-10UG CA76-50UG CA76-500UG CA76-1MG
102943-594EA 57.08 USD
102943-594 102943-596 102943-598 102943-600
Human Recombinant Activin Receptor Type-1
Proteins and Peptides
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels
Recombinant Human Activin Receptor IA is produced by our Mammalian expression system and the target gene encoding Met21-Val124 is expressed with a 6His tag at the C-terminus.
Información accesorios: Accessories not included.
Información de suministro: Shipped Via FedEx
Envase: Shipped at ambient temperature via FedEx envelope.
Precaución: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.