Human Recombinant Lymphocyte Antigen 6H
Supplier: Bon Opus Biosciences
CA64-10UG CA64-50UG CA64-500UG CA64-1MG
102943-570EA 170.15 USD
102943-570 102943-572 102943-574 102943-576
Human Recombinant Lymphocyte Antigen 6H
Proteins and Peptides
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels
Recombinant Human Lymphocyte Antigen 6H is produced by our Mammalian expression system and the target gene encoding Leu26-Gly115 is expressed with a 6His tag at the C-terminus.
Información accesorios: Accessories not included.
Información de suministro: Shipped Via FedEx
Envase: Shipped at ambient temperature via FedEx envelope.
Precaución: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.