Human Recombinant ERO1-Like Protein Alpha
Supplier: Bon Opus Biosciences
CA42-10UG CA42-50UG CA42-500UG CA42-1MG
102943-514EA 170.15 USD
102943-514 102943-516 102943-518 102943-520
Human Recombinant ERO1-Like Protein Alpha
Proteins and Peptides
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels
Recombinant Human ERO1L is produced by our Mammalian expression system and the target gene encoding Glu24-His468 is expressed with a 6His tag at the C-terminus.
Información accesorios: Accessories not included.
Información de suministro: Shipped Via FedEx
Envase: Shipped at ambient temperature via FedEx envelope.
Precaución: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.