Human Recombinant Carboxypeptidase B2
Supplier: Bon Opus Biosciences
CA32-10UG CA32-50UG CA32-500UG CA32-1MG
102943-474EA 118.46 USD
102943-474 102943-476 102943-478 102943-480
Human Recombinant Carboxypeptidase B2
Proteins and Peptides
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels
Recombinant Human Carboxypeptidase B2 is produced by our Mammalian expression system and the target gene encoding Phe23-Val423 is expressed with a 6His tag at the C-terminus.
Información accesorios: Accessories not included.
Información de suministro: Shipped Via FedEx
Envase: Shipped domestic on polar packs in a 9"x6"x7" box, or shipped internationally on Dry Ice in a 14" cube box.
Precaución: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.