Human Recombinant Protocadherin-1
Supplier: Bon Opus Biosciences
C844-10UG C844-50UG C844-500UG C844-1MG
102943-138EA 170.15 USD
102943-138 102943-140 102943-142 102943-144
Human Recombinant Protocadherin-1
Proteins and Peptides
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels
Recombinant Human Protocadherin-1 is produced by our Mammalian expression system and the target gene encoding Thr58-Asn852 is expressed with a 6His tag at the C-terminus.
Información accesorios: Accessories not included.
Información de suministro: Shipped Via FedEx
Envase: Shipped at ambient temperature via FedEx envelope.
Precaución: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.