Human Recombinant Carbonic Anhydrase 11
Supplier: Bon Opus Biosciences
C823-10UG C823-50UG C823-500UG C823-1MG
102943-090EA 170.15 USD
102943-090 102943-092 102943-094 102943-096
Human Recombinant Carbonic Anhydrase 11
Enzymes
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels
Recombinant Human Carbonic Anhydrase 11 is produced by our Mammalian expression system and the target gene encoding His24-Arg328 is expressed with a 6His tag at the C-terminus.
Información accesorios: Accessories not included.
Información de suministro: Shipped Via FedEx
Envase: Shipped domestic on polar packs in a 9×6×7" box, or shipped internationally on Dry Ice in a 14" cube box.
Precaución: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.