Human Recombinant Interleukin-25
Supplier: Bon Opus Biosciences
C792-10UG C792-50UG C792-500UG C792-1MG
102942-978EA 118.46 USD
102942-978 102942-980 102942-982 102942-984
Human Recombinant Interleukin-25
Proteins and Peptides
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels
Recombinant Human Interleukin-25 is produced by our Mammalian expression system and the target gene encoding Tyr33-Gly177 is expressed with a 6His tag at the C-terminus.
Información accesorios: Accessories not included.
Información de suministro: Shipped Via FedEx
Envase: Shipped at ambient temperature via FedEx envelope.
Precaución: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.