Mouse Recombinant Growth-regulated alpha protein
Supplier: Bon Opus Biosciences
C786-10UG C786-50UG C786-500UG C786-1MG
102942-930EA 118.46 USD
102942-930 102942-932 102942-934 102942-936
Mouse Recombinant Growth-regulated alpha protein
Proteins and Peptides
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels
Recombinant Mouse C-X-C motif chemokine 1 is produced by our Mammalian expression system and the target gene encoding Arg20-Lys96 is expressed with a 6His tag at the C-terminus.
Información accesorios: Accessories not included.
Información de suministro: Shipped Via FedEx
Envase: Shipped at ambient temperature via FedEx envelope.
Precaución: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.