Recombinant Interferon lambda-1
Supplier: Bon Opus Biosciences
C668-10UG C668-50UG C668-500UG C668-1MG
103529-244EA 85.08 USD
103529-244 103529-278 103529-352 103529-402
Recombinant Interferon lambda-1
Proteins and Peptides
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels
Recombinant Human Interferon Lambda-1 is produced by our Mammalian expression system and the target gene encoding Gly20-Thr200 is expressed with a 6His at the C-terminus.
Información accesorios: Accessories not included.
Información de suministro: Shipped Via FedEx
Envase: Shipped at ambient temperature via FedEx envelope.
Precaución: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.