Recombinant Human Nogo Receptor/Ngr, Bon Opus Biosciences
Supplier: Bon Opus Biosciences
C632-10UG C632-1MG C632-500UG C632-50UG
102863-294EA 79.69 USD
102863-294 102863-296 102863-298 102863-300
Recombinant Human Nogo Receptor/Ngr, Bon Opus Biosciences
Proteins and Peptides
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels
Recombinant Human NgR is produced by our Mammalian expression system and the target gene encoding Cys27-Ser447 is expressed with a 6His tag at the C-terminus.
Información accesorios: Accessories not included.
Información de suministro: Shipped Via FedEx
Envase: Shipped at ambient temperature via FedEx envelope.
Precaución: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.