Recombinant Human Nectin-3, Bon Opus Biosciences
Supplier: Bon Opus Biosciences
C630-10UG C630-1MG C630-500UG C630-50UG
102863-286EA 90.46 USD
102863-286 102863-288 102863-290 102863-292
Recombinant Human Nectin-3, Bon Opus Biosciences
Proteins and Peptides
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels
Recombinant Human Nectin-3 is produced by our Mammalian expression system and the target gene encoding Gly58-Cys366 is expressed with a 6His tag at the C-terminus.
Información accesorios: Accessories not included.
Información de suministro: Shipped Via FedEx
Envase: Shipped at ambient temperature via FedEx envelope.
Precaución: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.