Recomb Human Transthyretin/Attr, Bon Opus Biosciences
Supplier: Bon Opus Biosciences
C545-10UG C545-1MG C545-500UG C545-50UG
102862-918EA 132.46 USD
102862-918 102862-920 102862-922 102862-924
Recomb Human Transthyretin/Attr, Bon Opus Biosciences
Proteins and Peptides
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels
Recombinant Human Transthyretin is produced by our Mammalian expression system and the target gene encoding Gly21-Glu147 is expressed with a 6His tag at the C-terminus.
Información accesorios: Accessories not included.
Información de suministro: Shipped Via FedEx
Envase: Shipped at ambient temperature via FedEx envelope.
Precaución: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.