Human Recombinant Pancreatic Secretory Trypsin Inhibitor
Supplier: Bon Opus Biosciences
C542-10UG C542-1MG C542-500UG C542-50UG
102862-894EA 118.46 USD
102862-894 102862-896 102862-898 102862-900
Human Recombinant Pancreatic Secretory Trypsin Inhibitor
Proteins and Peptides
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels
Recombinant Human SPINK1 is produced by our Mammalian expression system and the target gene encoding Asp24-Cys79 is expressed with a 6His tag at the C-terminus.
Información accesorios: Accessories not included.
Información de suministro: Shipped Via FedEx
Envase: Shipped domestic on polar packs in a 9"x6"x7" box, or shipped internationally on Dry Ice in a 14" cube box.
Precaución: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.