Human Recombinant Plasma Protease C1 Inhibitor
Supplier: Bon Opus Biosciences
C539-10UG C539-1MG C539-500UG C539-50UG
102862-870EA 118.46 USD
102862-870 102862-872 102862-874 102862-876
Human Recombinant Plasma Protease C1 Inhibitor
Proteins and Peptides
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels
Recombinant Human Serpin G1 is produced by our Mammalian expression system and the target gene encoding Asn23-Ala500 is expressed with a 6His tag at the C-terminus.
Información accesorios: Accessories not included.
Información de suministro: Shipped Via FedEx
Envase: Shipped at ambient temperature via FedEx envelope.
Precaución: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.