Human Recombinant Plasminogen Activator Inhibitor 1
Supplier: Bon Opus Biosciences
C537-10UG C537-1MG C537-500UG C537-50UG
102862-854EA 170.15 USD
102862-854 102862-856 102862-858 102862-860
Human Recombinant Plasminogen Activator Inhibitor 1
Proteins and Peptides
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels
Recombinant Human Serpin E1/PAI-1 is produced by our Mammalian expression system and the target gene encoding Val24-Pro402 is expressed with a 6His tag at the C-terminus.
Información accesorios: Accessories not included.
Información de suministro: Shipped Via FedEx
Envase: Shipped at ambient temperature via FedEx envelope.
Precaución: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.