Human Recombinant Lysosome Membrane Protein 2
Supplier: Bon Opus Biosciences
C532-10UG C532-1MG C532-500UG C532-50UG
102862-814EA 144.31 USD
102862-814 102862-816 102862-818 102862-820
Human Recombinant Lysosome Membrane Protein 2
Proteins and Peptides
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels
Recombinant Human SCARB2 is produced by our Mammalian expression system and the target gene encoding Arg27-Thr432 is expressed with a 6His tag at the C-terminus.
Información accesorios: Accessories not included.
Información de suministro: Shipped Via FedEx
Envase: Shipped at ambient temperature via FedEx envelope.
Precaución: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.