Human Recombinant Tie-1
Supplier: Bon Opus Biosciences
C502-10UG C502-1MG C502-500UG C502-50UG
102862-606EA 45.23 USD
102862-606 102862-608 102862-610 102862-612
Human Recombinant Tie-1
Enzymes
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels
Recombinant Human Tie-1 is produced by our Mammalian expression system and the target gene encoding Ala22-Gln760 is expressed with a 6His tag at the C-terminus.
Información accesorios: Accessories not included.
Información de suministro: Shipped Via FedEx
Envase: Shipped at ambient temperature via FedEx envelope.
Precaución: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.