Human Recombinant Leukocyte Immunoglobulin-Like Receptor Subfamily B Member 2
Supplier: Bon Opus Biosciences
C485-10UG C485-1MG C485-500UG C485-50UG
102862-486EA 79.69 USD
102862-486 102862-488 102862-490 102862-492
Human Recombinant Leukocyte Immunoglobulin-Like Receptor Subfamily B Member 2
Proteins and Peptides
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels
Recombinant Human LILRB2 is produced by our Mammalian expression system and the target gene encoding Gln22-His458 is expressed with a 6His tag at the C-terminus.
Información accesorios: Accessories not included.
Información de suministro: Shipped Via FedEx
Envase: Shipped at ambient temperature via FedEx envelope.
Precaución: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.