Human Recombinant Junctional Adhesion Molecule A
Supplier: Bon Opus Biosciences
C468-10UG C468-1MG C468-500UG C468-50UG
102862-366EA 118.46 USD
102862-366 102862-368 102862-370 102862-372
Human Recombinant Junctional Adhesion Molecule A
Proteins and Peptides
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels
Recombinant Human JAM-A is produced by our Mammalian expression system and the target gene encoding Ser28-Val238 is expressed with a 6His tag at the C-terminus.
Información accesorios: Accessories not included.
Información de suministro: Shipped Via FedEx
Envase: Shipped at ambient temperature via FedEx envelope.
Precaución: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.