Human Recombinant 5'-Nucleotidase
Supplier: Bon Opus Biosciences
C446-10UG C446-1MG C446-500UG C446-50UG
102862-198EA 170.15 USD
102862-198 102862-200 102862-202 102862-204
Human Recombinant 5'-Nucleotidase
Proteins and Peptides
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels
Recombinant Human 5'-Nucleotidase is produced by our Mammalian expression system and the target gene encoding Trp27-Lys547 is expressed with a 6His tag at the C-terminus.
Información accesorios: Accessories not included.
Información de suministro: Shipped Via FedEx
Envase: Shipped domestic on polar packs in a 9"x6"x7" box, or shipped internationally on Dry Ice in a 14" cube box.
Precaución: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.