Human Recombinant Myeloid Cell Surface Antigen CD33
Supplier: Bon Opus Biosciences
C445-10UG C445-1MG C445-500UG C445-50UG
102862-190EA 90.46 USD
102862-190 102862-192 102862-194 102862-196
Human Recombinant Myeloid Cell Surface Antigen CD33
Proteins and Peptides
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels
Recombinant Human Sialic Acid Binding Ig-like Lectin 3 is produced by our Mammalian expression system and the target gene encoding Asp18-His259 is expressed with a Fc, 6His tag at the C-terminus.
Información accesorios: Accessories not included.
Información de suministro: Shipped Via FedEx
Envase: Shipped at ambient temperature via FedEx envelope.
Precaución: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.