Human Recombinant Cathepsin D
Supplier: Bon Opus Biosciences
C399-10UG C399-1MG C399-500UG C399-50UG
102861-886EA 170.15 USD
102861-886 102861-888 102861-890 102861-892
Human Recombinant Cathepsin D
Proteins and Peptides
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels
Recombinant Human Cathepsin D is produced by our Mammalian expression system and the target gene encoding Leu21-Leu412 is expressed with a 6His tag at the C-terminus.
Información accesorios: Accessories not included.
Información de suministro: Shipped Via FedEx
Envase: Shipped domestic on polar packs in a 9"x6"x7" box, or shipped internationally on Dry Ice in a 14" cube box.
Precaución: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.