Human Recombinant Leucine-Rich Repeat Transmembrane Protein FLRT2
Supplier: Bon Opus Biosciences
C349-10UG C349-1MG C349-500UG C349-50UG
102861-534EA 90.46 USD
102861-534 102861-536 102861-538 102861-540
Human Recombinant Leucine-Rich Repeat Transmembrane Protein FLRT2
Proteins and Peptides
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels
Recombinant Human FLRT2 is produced by our Mammalian expression system and the target gene encoding Cys36-Ser539 is expressed with a 6His tag at the C-terminus.
Información accesorios: Accessories not included.
Información de suministro: Shipped Via FedEx
Envase: Shipped at ambient temperature via FedEx envelope.
Precaución: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.