Human Recombinant Protein FAM3C
Supplier: Bon Opus Biosciences
C343-10UG C343-1MG C343-500UG C343-50UG
102861-494EA 118.46 USD
102861-494 102861-496 102861-498 102861-500
Human Recombinant Protein FAM3C
Proteins and Peptides
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels
Recombinant Human Protein FAM3C is produced by our Mammalian expression system and the target gene encoding Gln25-Asp227 is expressed with a 6His tag at the C-terminus.
Información accesorios: Accessories not included.
Información de suministro: Shipped Via FedEx
Envase: Shipped at ambient temperature via FedEx envelope.
Precaución: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.