Human Recombinant Coxsackievirus and Adenovirus Receptor
Supplier: Bon Opus Biosciences
C332-10UG C332-1MG C332-500UG C332-50UG
102861-422EA 90.46 USD
102861-422 102861-424 102861-426 102861-428
Human Recombinant Coxsackievirus and Adenovirus Receptor
Proteins and Peptides
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels
Recombinant Human Coxsackievirus and Adenovirus Receptor is produced by our Mammalian expression system and the target gene encoding Leu20-Gly237 is expressed with a 6His tag at the C-terminus.
Información accesorios: Accessories not included.
Información de suministro: Shipped Via FedEx
Envase: Shipped at ambient temperature via FedEx envelope.
Precaución: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.