Human Recombinant Cytotoxic and Regulatory T-Cell Molecule
Supplier: Bon Opus Biosciences
C331-10UG C331-1MG C331-500UG C331-50UG
102861-414EA 90.46 USD
102861-414 102861-416 102861-418 102861-420
Human Recombinant Cytotoxic and Regulatory T-Cell Molecule
Proteins and Peptides
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels
Recombinant Human CD355 is produced by our Mammalian expression system and the target gene encoding Ser18-Ser286 is expressed with a 6His tag at the C-terminus.
Información accesorios: Accessories not included.
Información de suministro: Shipped Via FedEx
Envase: Shipped at ambient temperature via FedEx envelope.
Precaución: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.