Human Recombinant Histone Chaperone ASF1A
Supplier: Bon Opus Biosciences
C290-10UG C290-1MG C290-500UG C290-50UG
102861-126EA 170.15 USD
102861-126 102861-128 102861-130 102861-132
Human Recombinant Histone Chaperone ASF1A
Proteins and Peptides
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels
Recombinant Human Histone Chaperone ASF1A is produced by our E.coli expression system and the target gene encoding Met1-Met204 is expressed with a T7 tag at the N-terminus, 6His tag at the C-terminus.
Información accesorios: Accessories not included.
Información de suministro: Shipped Via FedEx
Envase: Shipped at ambient temperature via FedEx envelope.
Precaución: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.