Human Recombinant Galectin-1
Supplier: Bon Opus Biosciences
C285-10UG C285-1MG C285-500UG C285-50UG
102861-086EA 43.08 USD
102861-086 102861-088 102861-090 102861-092
Human Recombinant Galectin-1
Proteins and Peptides
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels
Recombinant Human Galectin-1 is produced by our E.coli expression system and the target gene encoding Ala2-Asp135 is expressed with a 6His tag at the C-terminus.
Información accesorios: Accessories not included.
Información de suministro: Shipped Via FedEx
Envase: Shipped at ambient temperature via FedEx envelope.
Precaución: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.