Recombinant Human Zmynd19/Mizip, Bon Opus Biosciences
Supplier: Bon Opus Biosciences
C280-10UG C280-1MG C280-500UG C280-50UG
102861-046EA 170.15 USD
102861-046 102861-048 102861-050 102861-052
Recombinant Human Zmynd19/Mizip, Bon Opus Biosciences
Proteins and Peptides
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels
Recombinant Human Zinc Finger MYND Domain-Containing Protein 19 is produced by our E.coli expression system and the target gene encoding Met1-Arg227 is expressed with a 6His tag at the N-terminus.
Información accesorios: Accessories not included.
Información de suministro: Shipped Via FedEx
Envase: Shipped at ambient temperature via FedEx envelope.
Precaución: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.