Human Recombinant Gamma-Aminobutyric Acid Receptor-Associated Protein-Like 1
Supplier: Bon Opus Biosciences
C269-10UG C269-1MG C269-500UG C269-50UG
102860-958EA 170.15 USD
102860-958 102860-960 102860-962 102860-964
Human Recombinant Gamma-Aminobutyric Acid Receptor-Associated Protein-Like 1
Proteins and Peptides
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels
Recombinant Human GABARAPL1 is produced by our E.coli expression system and the target gene encoding Met1-Lys117 is expressed with a 6His tag at the N-terminus.
Información accesorios: Accessories not included.
Información de suministro: Shipped Via FedEx
Envase: Shipped at ambient temperature via FedEx envelope.
Precaución: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.