Recomb Human Prefoldin Subunit 2, Bon Opus Biosciences
Supplier: Bon Opus Biosciences
C254-10UG C254-1MG C254-500UG C254-50UG
102860-838EA 170.15 USD
102860-838 102860-840 102860-842 102860-844
Recomb Human Prefoldin Subunit 2, Bon Opus Biosciences
Proteins and Peptides
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels
Recombinant Human Prefoldin Subunit 2 is produced by our E.coli expression system and the target gene encoding Met1-Ser154 is expressed with a 6His tag at the N-terminus.
Información accesorios: Accessories not included.
Información de suministro: Shipped Via FedEx
Envase: Shipped at ambient temperature via FedEx envelope.
Precaución: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.