Human Recombinant Deoxyhypusine Synthase (from E. coli)
Supplier: Bon Opus Biosciences
C129-10UG C129-1MG C129-500UG C129-50UG
102859-854EA 118.46 USD
102859-854 102859-856 102859-858 102859-860
Human Recombinant Deoxyhypusine Synthase (from E. coli)
Enzymes
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels
Recombinant Human Deoxyhypusine Synthase is produced by our E.coli expression system and the target gene encoding Met1-Asp369 is expressed with a 6His tag at the C-terminus.
Información accesorios: Accessories not included.
Información de suministro: Shipped Via FedEx
Envase: Shipped domestic on polar packs in a 9×6×7" box, or shipped internationally on Dry Ice in a 14" cube box.
Precaución: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.