Human Recombinant Carbonic Anhydrase 8
Supplier: Bon Opus Biosciences
C110-10UG C110-1MG C110-500UG C110-50UG
102859-702EA 170.15 USD
102859-702 102859-704 102859-706 102859-708
Human Recombinant Carbonic Anhydrase 8
Enzymes
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels
Recombinant Human Carbonic Anhydrase 5B is produced by our E.coli expression system and the target gene encoding Cys34-Pro317 is expressed with a 6His tag at the C-terminus.
Información accesorios: Accessories not included.
Información de suministro: Shipped Via FedEx
Envase: Shipped domestic on polar packs in a 9×6×7" box, or shipped internationally on Dry Ice in a 14" cube box.
Precaución: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.