Human Recombinant C-C Motif Chemokine 1
Supplier: Bon Opus Biosciences
C094-10UG C094-1MG C094-500UG C094-50UG
102859-574EA 170.15 USD
102859-574 102859-576 102859-578 102859-580
Human Recombinant C-C Motif Chemokine 1
Proteins and Peptides
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels
Recombinant Human C-C Motif Chemokine 1 is produced by our E.coli expression system and the target gene encoding Lys24-Lys96 is expressed.
Información accesorios: Accessories not included.
Información de suministro: Shipped Via FedEx
Envase: Shipped at ambient temperature via FedEx envelope.
Precaución: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.