Human Recombinant Interleukin-4
Supplier: Bon Opus Biosciences
C050-10UG C050-1MG C050-500UG C050-50UG
102859-294EA 132.46 USD
102859-294 102859-296 102859-298 102859-300
Human Recombinant Interleukin-4
Proteins and Peptides
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels
Recombinant Human Interleukin-4 is produced by our E.coli expression system and the target gene encoding His25-Ser153 is expressed.
Información accesorios: Accessories not included.
Información de suministro: Shipped Via FedEx
Envase: Shipped at ambient temperature via FedEx envelope.
Precaución: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.