Process QDS® Process and Research Cleaners
Supplier: Steris
Process QDS®
1J7501 1J7508
21899-730CS 365.16 USD
21899-730 14004-114
Process QDS® Process and Research Cleaners
Cleaning Agents Disinfectants
STERIS' Process QDS® Process and Research Cleaner is a liquid concentrate specifically formulated for the disinfection of hard non-porous surfaces. The use-dilution can be applied manually or sprayed..
  • Efficacy - Quaternary ammonium active ingredients provide broad spectrum antimicrobial efficacy
  • Productivity - Formulation includes surfactants for surface cleaning while disinfecting
  • Reliability - Ideal for use in a contamination control rotation
  • Flexibility - Can be used as a disinfectant or sanitizer
  • Assurance - Complete lot traceability with each lot documented by Certificate of Analysis
STERIS' Process QDS® Process and research cleaner is a phosphate-free, one-step cleaner/disinfectant formulated for use on hard non-porous surfaces. This product is recommended for use in disinfecting, sanitizing, and deodorizing hard non-porous surfaces. It is effective in hard water and organic soil loads.