Hydrofluoric acid 40 - 45%, Technical Grade, Fluka™

Supplier: Honeywell Research Chemicals
Danger

01066-5L 01066-6X1L
BJ01066-1LCS 323.06 USD
BJ01066-1L BJ01066-5L BJ01066-6X1L
Hydrofluoric acid 40 - 45%, Technical Grade, Fluka™
Hydrofluoric acid

  • Chemicals that do not have an established standard set for quality and impurity levels or for products where the purity is <90%
  • Variation in color and physical form is possible

Formula: HF
MW: 20.01
Boiling Pt: 110 °C (1.013 hPa)
Melting Pt: –40 °C
Density: 1.140 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011346
CAS Number: 7664-39-3
EINECS: 231-634-8
UN: 1790
ADR: 8 II

Order Now

Specification Test Results

Vapor Density 1.27 (vs air)
Assay 40 - 45 %
Hexafluorosilicic acid (H₂SiF₆) Max. 0.02 %
Sulfate (SO₄) Max. 0.01 %
Vapor Pressure 25 mmHg ( 20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR